AC5947208556 Accela ChemBio Inc.

N-Boc-L-glutamic Acid 1,5-Dimethyl Ester

  • MDL number MFCD03094774
  • Cas Number : 59279-60-6
  • Molecular Weight : 275.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY041419&attname=GoodsCode

Safety Information

hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY041419.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart