AC5947208563 Accela ChemBio Inc.

Methyl 2-Hydroxypyrimidine-5-carboxylate

  • MDL number MFCD11848117
  • Cas Number : 50628-34-7
  • Molecular Weight : 154.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY041468&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY041468.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart