AC5947208584 Accela ChemBio Inc.

2-Iodophenyl Isothiocyanate

  • MDL number MFCD00041057
  • Cas Number : 98041-44-2
  • Molecular Weight : 261.08283

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY041528&attname=GoodsCode

Safety Information

signalword Danger
pcodes P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501
hcodes GHS05,GHS06
MSDS http://pdf.accelachem.com/cn/msds/SY041528.pdf
hstatements H331-H302-H312-H314-H318
Availability details for
Quantity
  • Availability
  • Add to Cart