AC5947208627 Accela ChemBio Inc.

2-(4-Imidazolyl)acetonitrile

  • MDL number MFCD00070306
  • Cas Number : 18502-05-1
  • Molecular Weight : 107.1133

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY041793&attname=GoodsCode

Safety Information

signalword Danger
pcodes P261-P280-P301+P310+P330-P302+P352+P312+P361+P364-P304+P340+P311-P305+P351+P338+P337+P313
hcodes GHS06
MSDS http://pdf.accelachem.com/cn/msds/SY041793.pdf
hstatements H301+H311+H331-H315-H319
Availability details for
Quantity
  • Availability
  • Add to Cart