AC5947208728 Accela ChemBio Inc.

1-(tert-Butyldimethylsilyl)-2-propen-1-ol

  • MDL number MFCD19687250
  • Cas Number : 105875-75-0
  • Molecular Weight : 172.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. RT
Physical Form Colorless to yellow liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY043363&attname=GoodsCode

Safety Information

signalword Warning
pcodes P210-P280-P240-P303+P361+P353-P403+P235-P501
hcodes GHS02
MSDS http://pdf.accelachem.com/cn/msds/SY043363.pdf
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart