AC5947208744 Accela ChemBio Inc.

Ethyl 2-(3,5-Dimethoxyphenyl)acetate

  • MDL number MFCD11111879
  • Cas Number : 65976-77-4
  • Molecular Weight : 224.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY043382&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY043382.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart