AC5947208850 Accela ChemBio Inc.

1-(Methoxymethyl)-4-nitropyrazole

  • MDL number MFCD16345427
  • Cas Number : 199274-87-8
  • Molecular Weight : 157.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY045097&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY045097.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart