AC5947208864 Accela ChemBio Inc.

1-Cyclopentyl-2-propyn-1-ol

  • MDL number MFCD21143284
  • Cas Number : 112256-09-4
  • Molecular Weight : 124.18028

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY045239&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY045239.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart