AC5947208933 Accela ChemBio Inc.

3,6-Dichloro-4-hydroxypyridazine

  • MDL number MFCD04971276
  • Cas Number : 2779-81-9
  • Molecular Weight : 164.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY045542&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY045542.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart