AC5947208985 Accela ChemBio Inc.

5-Cyanobenzofuran-2-carboxylic Acid

  • MDL number MFCD09837162
  • Cas Number : 84102-75-0
  • Molecular Weight : 187.1516

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY045690&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY045690.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart