AC5947209003 Accela ChemBio Inc.

1-Fluoro-2-propylamine Hydrochloride

  • MDL number MFCD20489620
  • Cas Number : 921602-78-0
  • Molecular Weight : 113.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY045872&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY045872.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart