AC5947209288 Accela ChemBio Inc.

2,6-Dibromo-4-methylpyridine

  • MDL number MFCD11036225
  • Cas Number : 73112-16-0
  • Molecular Weight : 250.9186

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY047640&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY047640.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart