AC5947209396 Accela ChemBio Inc.

2-Methoxy-5-nitrophenylacetylene

  • MDL number MFCD11656154
  • Cas Number : 77123-62-7
  • Molecular Weight : 177.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to orange solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY048028&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY048028.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart