AC5947209507 Accela ChemBio Inc.

3,6-Dibromothieno[3,2-b]thiophene

  • MDL number MFCD10000958
  • Cas Number : 392662-65-6
  • Molecular Weight : 298.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY051420&attname=GoodsCode

Safety Information

hcodes no data known
MSDS http://pdf.accelachem.com/cn/msds/SY051420.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart