AC5947209563 Accela ChemBio Inc.

Methyl 3,5-Dinitrosalicylate

  • MDL number MFCD00017561
  • Cas Number : 22633-33-6
  • Molecular Weight : 242.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY056856&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY056856.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart