AC5947209595 Accela ChemBio Inc.

4-(Aminomethyl)-2-pyrimidinamine

  • MDL number MFCD09040794
  • Cas Number : 929973-95-5
  • Molecular Weight : 124.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY057525&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY057525.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart