AC5947209597 Accela ChemBio Inc.

(11beta,16alpha)-11,16,17,21-Tetrahydroxypregna-1,4-diene-3,20-dione

  • MDL number MFCD00872019
  • Cas Number : 13951-70-7
  • Molecular Weight : 376.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY057544&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY057544.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart