AC5947209676 Accela ChemBio Inc.

(4-Methoxyphenyl)(4-pyridyl)methanone

  • MDL number MFCD01716434
  • Cas Number : 14548-47-1
  • Molecular Weight : 213.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY058187&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY058187.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart