AC5947209747 Accela ChemBio Inc.

(S)-3-Acetoxy-4-ethoxy-4-oxobutyric Acid

  • MDL number MFCD30378926
  • Cas Number : 52485-05-9
  • Molecular Weight : 204.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY046161&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY046161.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart