AC5947209859 Accela ChemBio Inc.

5-Amino-2-(1,3-dioxoisoindolin-2-yl)-5-oxopentanoic Acid

  • MDL number MFCD00422111
  • Cas Number : 7607-72-9
  • Molecular Weight : 276.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY059199&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY059199.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart