AC5947209956 Accela ChemBio Inc.

3-Amino-5-(3,5-difluorobenzyl)-1H-indazole

  • MDL number MFCD28129098
  • Cas Number : 1108745-30-7
  • Molecular Weight : 259.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY060686&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY060686.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart