AC5998047344 Accela ChemBio Inc.

4-Methoxyindole-3-acetic Acid

  • MDL number MFCD10035274
  • Cas Number : 17897-49-3
  • Molecular Weight : 205.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY058615&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY058615.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart