AC5998047366 Accela ChemBio Inc.

Ethyl 6-Nitroindole-3-acetate

  • MDL number MFCD30742646
  • Cas Number : 501919-82-0
  • Molecular Weight : 248.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY058637&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY058637.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart