AC5998047466 Accela ChemBio Inc.

4-Dimethylaminoazobenzene-4'-carboxylic Acid

  • MDL number MFCD00058988
  • Cas Number : 6268-49-1
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form Yellow to reddish brown solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY059974&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY059974.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart