AC5998047492 Accela ChemBio Inc.

6-Bromo-2,2-dimethyl-2H-pyrido[3,2-b][1,4]oxazin-3(4H)-one

  • MDL number MFCD10000813
  • Cas Number : 1196153-28-2
  • Molecular Weight : 257.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY060528&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY060528.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart