AC5H03BB219C Accela ChemBio Inc.

4-[3-[4-(Cyclopropanecarbonyl)piperazine-1-carbonyl]-4-fluorobenzyl]phthalazin-1(2H)-one

  • MDL number MFCD13185161
  • Cas Number : 763113-22-0
  • Molecular Weight : 434.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY040527&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY040527.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart