AC5H03BB21BE Accela ChemBio Inc.

7-Bromo-3,4-dihydro-1,5-naphthyridin-2(1H)-one

  • MDL number MFCD18260394
  • Cas Number : 1256834-74-8
  • Molecular Weight : 227.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY060537&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY060537.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart