AC5H03BB21E2 Accela ChemBio Inc.

1-Boc-piperidine-2-boronic Acid Pinacol Ester

  • MDL number MFCD24476416
  • Cas Number : 2071192-58-8
  • Molecular Weight : 311.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY122658&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY122658.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart