AC5H03BB21F2 Accela ChemBio Inc.

2-Bromo-5-(2-fluoro-2-propyl)pyridine

  • MDL number MFCD30012386
  • Cas Number : 2091683-62-2
  • Molecular Weight : 218.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY128733&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY128733.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart