AC5H04252A79 Accela ChemBio Inc.

Allylchloro[1,3-bis(2,6-di-i-propylphenyl)imidazol-2-ylidene]palladium(II)

  • MDL number MFCD08561131
  • Cas Number : 478980-03-9
  • Molecular Weight : 571.54

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name allyl(1,3-bis(2,6-diisopropylphenyl)-1,3-dihydro-2H-imidazol-2-ylidene)palladium(IV) chloride
Inchi Code 1S/C27H36N2.C3H5.ClH.Pd/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;1-3-2;;/h9-16,18-21H,1-8H3;3H,1-2H2;1H;/q;;;+1/p-1
Inchi Key SCISXDQUNWTGKI-UHFFFAOYSA-M

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY075811&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P305+P351+P338
MSDS http://pdf.accelachem.com/cn/msds/SY075811.pdf
hcodes GHS07
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart