AC5H0A313768 Accela ChemBio Inc.

4-(4-Morpholino-1-piperidinyl)aniline

  • MDL number MFCD20257438
  • Cas Number : 867291-42-7
  • Molecular Weight : 261.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form Yellow to tan solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY225344&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY225344.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart