AC5H0A313785 Accela ChemBio Inc.

Ethyl (E)-3-[4-(Benzyloxy)-3,5-dimethoxyphenyl]acrylate

  • MDL number MFCD31619545
  • Cas Number : 178611-06-8
  • Molecular Weight : 342.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226393&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226393.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart