AC5H0D73F7AB Accela ChemBio Inc.

4-Aminobenzoxazole

  • MDL number MFCD00973373
  • Cas Number : 163808-09-1
  • Molecular Weight : 134.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY012212&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY012212.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart