AC5H0D73F7CF Accela ChemBio Inc.

2,3-Dimethoxyaniline

  • MDL number MFCD00180769
  • Cas Number : 6299-67-8
  • Molecular Weight : 153.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. +4?
Physical Form Colorless to brown liquid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY030638&attname=GoodsCode

Safety Information

signalword Warning
pcodes P305 + P351 + P338
MSDS http://pdf.accelachem.com/cn/msds/SY030638.pdf
pictograms GHS07
hstatements H302-H319
Availability details for
Quantity
  • Availability
  • Add to Cart