AC5H0D73F7E0 Accela ChemBio Inc.

Benzyl 1-Boc-piperidine-3-carboxylate

  • MDL number MFCD11975554
  • Cas Number : 139985-95-8
  • Molecular Weight : 319.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY044918&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY044918.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart