AC5H0D73F7E6 Accela ChemBio Inc.

4,4-Dimethylcyclohexanol

  • MDL number MFCD00101954
  • Cas Number : 932-01-4
  • Molecular Weight : 128.21204

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY047599&attname=GoodsCode

Safety Information

signalword Warning
pcodes P280-P305+P351+P338-P304+P340-P302+P352-P312
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY047599.pdf
hstatements H319-H315
Availability details for
Quantity
  • Availability
  • Add to Cart