AC5H0D73F7E8 Accela ChemBio Inc.

1-Hydroxyanthraquinone

  • MDL number MFCD00058946
  • Cas Number : 129-43-1
  • Molecular Weight : 224.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form White to red solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY052323&attname=GoodsCode

Safety Information

signalword Warning
pcodes P201-P264-P280-P305+P351+P338+P337+P313-P308+P313-P405
MSDS http://pdf.accelachem.com/cn/msds/SY052323.pdf
pictograms GHS07,GHS08
hstatements H319-H351
Availability details for
Quantity
  • Availability
  • Add to Cart