AC5H0D73F7F8 Accela ChemBio Inc.

Ethyl 6-Formylnicotinate

  • MDL number MFCD19690524
  • Cas Number : 20857-31-2
  • Molecular Weight : 179.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY116615&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY116615.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart