AC5H0D73F801 Accela ChemBio Inc.

5-(Hydroxymethyl)pyridine-2-carboxaldehyde

  • MDL number MFCD18384433
  • Cas Number : 40749-33-5
  • Molecular Weight : 137.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY225996&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY225996.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart