AC5H0D73F802 Accela ChemBio Inc.

5-Methyloxazole-4-acetic Acid

  • MDL number MFCD31630346
  • Cas Number : 1507656-31-6
  • Molecular Weight : 141.12468

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226060&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226060.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart