AC5H0D73F807 Accela ChemBio Inc.

1,2,4-Tribromo-5-fluorobenzene

  • MDL number MFCD18089318
  • Cas Number : 7655-70-1
  • Molecular Weight : 332.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226345&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226345.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart