AC5H0D73F80B Accela ChemBio Inc.

1-Cyclopropyl-2-propyn-1-amine

  • MDL number MFCD21263287
  • Cas Number : 1462237-17-7
  • Molecular Weight : 95.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226575&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226575.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart