AC5H0D73F80D Accela ChemBio Inc.

1-[4-(Trifluoromethyl)-2-pyridyl]piperidine-4-carboxylic Acid

  • MDL number MFCD18256630
  • Cas Number : 1256792-04-7
  • Molecular Weight : 274.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226719&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226719.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart