AC5H0D73F812 Accela ChemBio Inc.

5,7-Dimethyl-2,3-dihydro-1,8-naphthyridin-4(1H)-one

  • MDL number MFCD18260807
  • Cas Number : 698973-85-2
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226809&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226809.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart