AC5H0D73F814 Accela ChemBio Inc.

Methyl 5-(Bromomethyl)-2-nitrobenzoate

  • MDL number MFCD20528191
  • Cas Number : 88071-91-4
  • Molecular Weight : 274.06812

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY226903&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY226903.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart