AC5H0D73F815 Accela ChemBio Inc.

1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea

  • MDL number MFCD00954637
  • Cas Number : 182498-32-4
  • Molecular Weight : 352.1402

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form solid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY231254&attname=GoodsCode

Safety Information

signalword Warning
pcodes P261-P280a-P305+P351+P338-P304+P340-P405-P501a
hcodes GHS07
MSDS http://pdf.accelachem.com/cn/msds/SY231254.pdf
hstatements H315-H319-H335
Availability details for
Quantity
  • Availability
  • Add to Cart