AC5H0D73F816 Accela ChemBio Inc.

Methyl 2-(3-Bromopropoxy)acetate

  • MDL number MFCD24459657
  • Cas Number : 433213-11-7
  • Molecular Weight : 211.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY231297&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY231297.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart