AC5H0D73F81A Accela ChemBio Inc.

4-(Cbz-amino)-1-methyl-1H-pyrazole-3-carboxylic Acid

  • MDL number MFCD31630369
  • Molecular Weight : 275.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4?
Physical Form White to yellow solid
Shipping Temperature RT
Country of Origin CN
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232208&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232208.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart