AC5H0D73F821 Accela ChemBio Inc.

Phenyl(tetrahydro-2H-pyran-4-yl)methanol

  • MDL number MFCD17243925
  • Cas Number : 216087-92-2
  • Molecular Weight : 192.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. +4?
Physical Form liquid
Shipping Temperature +4?
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY232891&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY232891.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart