AC5H30533A76 Accela ChemBio Inc.

1-(2,4-Dichlorophenyl)-2-(1-imidazolyl)ethanol

  • MDL number MFCD00044708
  • Cas Number : 24155-42-8
  • Molecular Weight : 257.1159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin China
Product Link http://www.accelachem.com/cn/search.html?keyword=SY002118&attname=GoodsCode

Safety Information

MSDS http://pdf.accelachem.com/cn/msds/SY002118.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart